Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001824.hg.1 |
TC6_mcf_hap5000099.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001824.hg.1 |
NM_030903.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000784.hg.1 |
TC6_apd_hap1000055.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000784.hg.1 |
NM_030903.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002082.hg.1 |
TC6_cox_hap2000117.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002082.hg.1 |
NM_030903.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016598.hg.1 |
TC06001480.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016598.hg.1 |
ENST00000377175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016598.hg.1 |
NM_030903.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002019.hg.1 |
TC6_dbb_hap3000105.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002019.hg.1 |
NM_030903.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011291.hg.1 |
ENST00000377175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011291.hg.1 |
ENST00000377175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001603.hg.1 |
TC6_mann_hap4000095.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001603.hg.1 |
NM_030903.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002064.hg.1 |
TC6_qbl_hap6000108.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002064.hg.1 |
NM_030903.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7001913.hg.1 |
TC6_ssto_hap7000099.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7001913.hg.1 |
NM_030903.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600176094.hg.1 |
ENST00000377175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176094.hg.1 |
ENST00000377175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176094.hg.1 |
TC0600011291.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600011291.hg.1 |
ENST00000377175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011291.hg.1 |
ENST00000377175.1 |
Ensembl |