Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
ENST00000342032.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
ENST00000342032.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
ENST00000342032.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
ENST00000342032.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
ENST00000342032.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
ENST00000342032.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
ENST00000342032.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
ENST00000342032.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
ENST00000342032.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
ENST00000342032.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
ENST00000342032.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
ENST00000342032.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
ENST00000409146.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
ENST00000409146.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
ENST00000409146.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
ENST00000409146.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
ENST00000409146.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
ENST00000409146.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
ENST00000498609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
ENST00000498609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
ENST00000498609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
ENST00000498609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
ENST00000498609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
NM_001145123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
NM_001145123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
NM_001145123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
NM_001145123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
NM_001145123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
NM_001145123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
NM_001145123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
NM_001145123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
NM_001145123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
NM_001145123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
NM_006658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
NM_006658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
NM_006658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
NM_006658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
NM_006658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
NM_006658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
NM_006658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
NM_006658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
NM_006658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
NM_006658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007112.hg.1 |
NM_006658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07003250.hg.1 |
TC07000204.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07003250.hg.1 |
ENST00000342032.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07003250.hg.1 |
ENST00000409146.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07003250.hg.1 |
NM_001145123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07003250.hg.1 |
NM_006658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07003251.hg.1 |
TC07000204.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07003251.hg.1 |
ENST00000342032.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07003251.hg.1 |
ENST00000409146.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07003251.hg.1 |
NM_001145123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07003251.hg.1 |
NM_006658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07003248.hg.1 |
TC07000204.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07003248.hg.1 |
ENST00000342032.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07003248.hg.1 |
NM_001145123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07003248.hg.1 |
NM_006658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700149515.hg.1 |
ENST00000342032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700149515.hg.1 |
ENST00000409146.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700149515.hg.1 |
NM_001145123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700149515.hg.1 |
NM_006658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700149515.hg.1 |
ENST00000342032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700149515.hg.1 |
ENST00000409146.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700149515.hg.1 |
NM_001145123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700149515.hg.1 |
NM_006658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700149515.hg.1 |
TC0700007112.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700149512.hg.1 |
ENST00000342032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700149512.hg.1 |
NM_001145123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700149512.hg.1 |
NM_006658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700149512.hg.1 |
ENST00000342032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700149512.hg.1 |
NM_001145123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700149512.hg.1 |
NM_006658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700149512.hg.1 |
TC0700007112.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700149514.hg.1 |
ENST00000342032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700149514.hg.1 |
ENST00000409146.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700149514.hg.1 |
NM_001145123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700149514.hg.1 |
NM_006658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700149514.hg.1 |
ENST00000342032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700149514.hg.1 |
ENST00000409146.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700149514.hg.1 |
NM_001145123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700149514.hg.1 |
NM_006658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700149514.hg.1 |
TC0700007112.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700149516.hg.1 |
ENST00000342032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700149516.hg.1 |
ENST00000409146.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700149516.hg.1 |
NM_001145123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700149516.hg.1 |
NM_006658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700149516.hg.1 |
ENST00000342032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700149516.hg.1 |
ENST00000409146.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700149516.hg.1 |
NM_001145123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700149516.hg.1 |
NM_006658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700149516.hg.1 |
TC0700007112.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
ENST00000342032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
ENST00000342032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
ENST00000342032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
ENST00000342032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
ENST00000342032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
ENST00000342032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
ENST00000342032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
ENST00000342032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
ENST00000342032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
ENST00000342032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
ENST00000342032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
ENST00000342032.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
ENST00000409146.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
ENST00000409146.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
ENST00000409146.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
ENST00000409146.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
ENST00000409146.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
ENST00000409146.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
ENST00000498609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
ENST00000498609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
ENST00000498609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
ENST00000498609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
ENST00000498609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
NM_001145123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
NM_001145123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
NM_001145123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
NM_001145123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
NM_001145123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
NM_001145123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
NM_001145123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
NM_001145123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
NM_001145123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
NM_001145123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
NM_006658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
NM_006658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
NM_006658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
NM_006658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
NM_006658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
NM_006658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
NM_006658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
NM_006658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
NM_006658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
NM_006658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007112.hg.1 |
NM_006658.4 |
Refseq |