Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022509.hg.1 |
TC05001664.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022509.hg.1 |
ENST00000361189.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022509.hg.1 |
ENST00000361557.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022509.hg.1 |
ENST00000511624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022509.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000361189.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000361189.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000361189.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000361189.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000361189.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000361189.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000361189.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000361189.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000361189.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000361189.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000361189.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000361189.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000361189.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000361189.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000361557.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000361557.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000361557.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000361557.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000361557.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000361557.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000361557.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000361557.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000361557.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000361557.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000361557.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000511624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000511624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000511624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000511624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000511624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000512822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000512822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000512822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000512822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
ENST00000512822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011672.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022508.hg.1 |
TC05001664.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022508.hg.1 |
ENST00000361189.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022508.hg.1 |
ENST00000361557.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022508.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500180706.hg.1 |
ENST00000361189.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180706.hg.1 |
ENST00000361557.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180706.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500180706.hg.1 |
ENST00000361189.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180706.hg.1 |
ENST00000361557.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180706.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500180706.hg.1 |
TC0500011672.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500180707.hg.1 |
ENST00000361189.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180707.hg.1 |
ENST00000361557.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180707.hg.1 |
ENST00000511624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180707.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500180707.hg.1 |
ENST00000361189.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180707.hg.1 |
ENST00000361557.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180707.hg.1 |
ENST00000511624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180707.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500180707.hg.1 |
TC0500011672.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000361189.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000361189.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000361189.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000361189.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000361189.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000361189.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000361189.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000361189.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000361189.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000361189.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000361189.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000361189.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000361189.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000361189.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000361557.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000361557.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000361557.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000361557.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000361557.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000361557.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000361557.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000361557.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000361557.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000361557.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000361557.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000511624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000511624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000511624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000511624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000511624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000512822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000512822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000512822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000512822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
ENST00000512822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
NM_014819.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011672.hg.1 |
NM_014819.4 |
Refseq |