Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027946.hg.1 |
ENST00000292095.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027946.hg.1 |
ENST00000313005.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027946.hg.1 |
ENST00000392937.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027946.hg.1 |
ENST00000428381.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027946.hg.1 |
ENST00000445823.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027946.hg.1 |
ENST00000510630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027946.hg.1 |
ENST00000510915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027946.hg.1 |
ENST00000513780.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027946.hg.1 |
ENST00000528053.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027946.hg.1 |
NM_001207048.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027946.hg.1 |
NM_001207049.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027946.hg.1 |
NM_012104.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027946.hg.1 |
NM_138971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027946.hg.1 |
NM_138972.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027946.hg.1 |
NM_138973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027946.hg.1 |
TC11002335.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
ENST00000313005.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
ENST00000392937.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
ENST00000428381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
ENST00000445823.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
ENST00000504995.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
ENST00000510630.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
ENST00000510915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
ENST00000513780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
ENST00000514464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
ENST00000528053.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
ENST00000530824.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
NM_001207048.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
NM_001207049.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
NM_012104.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
NM_138971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
NM_138972.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
NM_138973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
ENST00000313005.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
ENST00000392937.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
ENST00000428381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
ENST00000445823.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
ENST00000504995.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
ENST00000510630.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
ENST00000510915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
ENST00000513780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
ENST00000514464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
ENST00000528053.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
ENST00000530824.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
NM_001207048.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
NM_001207049.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
NM_012104.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
NM_138971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
NM_138972.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
NM_138973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195597.hg.1 |
TC1100012445.hg.1 |
Clariom D TC |