Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007141.hg.1 |
TC05000515.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007141.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007141.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000502261.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000502261.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000502261.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000503970.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000503970.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000503970.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000503970.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000503970.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000503970.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000505313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000505313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000505313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000505313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000508043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000508043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000508043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000508043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000513921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000513921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000513921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000513921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000513921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
ENST00000513921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500156735.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500156735.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500156735.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500156735.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500156735.hg.1 |
TC0500008296.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000261483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000502261.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000502261.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000502261.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000503970.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000503970.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000503970.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000503970.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000503970.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000503970.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000505313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000505313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000505313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000505313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000508043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000508043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000508043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000508043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000513921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000513921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000513921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000513921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000513921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
ENST00000513921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008296.hg.1 |
NM_002372.3 |
Refseq |