Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000477934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000477934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000477934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000477934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000483388.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000483388.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000483388.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000483388.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000483388.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000483388.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000483388.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000490959.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000490959.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000490959.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000490959.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007716.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006836.hg.1 |
TC01000440.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006836.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006836.hg.1 |
ENST00000477934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006836.hg.1 |
ENST00000539719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006836.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006837.hg.1 |
TC01000440.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006837.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006837.hg.1 |
ENST00000477934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006837.hg.1 |
ENST00000539719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006837.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006838.hg.1 |
TC01000440.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006838.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006838.hg.1 |
ENST00000477934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006838.hg.1 |
ENST00000539719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006838.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100154331.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154331.hg.1 |
ENST00000477934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154331.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154331.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100154331.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154331.hg.1 |
ENST00000477934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154331.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154331.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100154331.hg.1 |
TC0100007716.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100154332.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154332.hg.1 |
ENST00000477934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154332.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154332.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100154332.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154332.hg.1 |
ENST00000477934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154332.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154332.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100154332.hg.1 |
TC0100007716.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000477934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000477934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000477934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000477934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000483388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000483388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000483388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000483388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000483388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000483388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000483388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000490959.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000490959.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000490959.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000490959.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007716.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100154330.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154330.hg.1 |
ENST00000477934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154330.hg.1 |
ENST00000483388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154330.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154330.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100154330.hg.1 |
ENST00000373428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154330.hg.1 |
ENST00000477934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154330.hg.1 |
ENST00000483388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154330.hg.1 |
ENST00000539719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154330.hg.1 |
NM_152493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100154330.hg.1 |
TC0100007716.hg.1 |
Clariom D TC |