Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000426.3 | NP_000417.2 | 55 - 56 | 67 | ||
NM_001079823.1 | NP_001073291.1 | 54 - 55 | 67 | |||
XM_005266981.3 | XP_005267038.1 | 56 - 57 | 8119 | 67 | ||
XM_005266982.3 | XP_005267039.1 | 55 - 56 | 8107 | 67 | ||
XM_011535820.2 | XP_011534122.1 | 55 - 56 | 8113 | 67 | ||
XM_017010851.1 | XP_016866340.1 | 56 - 57 | 10507 | 67 | ||
XM_017010852.1 | XP_016866341.1 | 45 - 46 | 9396 | 67 | ||
GenBank mRNA | AB208922.1 | - | 29 - 30 | 3945 | 67 | |
M59832.1 | - | 15 - 16 | 67 | |||
Z26653.1 | - | 55 - 56 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |