Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032471.5 | NP_115860.1 | 2 - 3 | 381 | 120 | |
GenBank mRNA | BC036011.1 | - | 2 - 3 | 213 | 4768945 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |