Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022449.hg.1 |
ENST00000291294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022449.hg.1 |
NM_000960.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022449.hg.1 |
TC19001650.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000291294.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000291294.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000291294.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000291294.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000291294.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000291294.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000291294.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000594275.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000594275.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000594275.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000594275.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000595460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000595460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000595460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000595460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000595460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000595460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000595460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000596260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000596260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000596260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000596260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000596260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000597185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000597185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000597185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000597185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000597185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000598865.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000598865.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000598865.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000598865.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
ENST00000598865.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
NM_000960.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
NM_000960.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
NM_000960.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
NM_000960.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
NM_000960.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
NM_000960.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010994.hg.1 |
NM_000960.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022450.hg.1 |
ENST00000291294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022450.hg.1 |
NM_000960.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022450.hg.1 |
TC19001650.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900191501.hg.1 |
ENST00000291294.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191501.hg.1 |
ENST00000595460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191501.hg.1 |
ENST00000596260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191501.hg.1 |
NM_000960.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900191501.hg.1 |
TC1900010994.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900191501.hg.1 |
ENST00000291294.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191501.hg.1 |
ENST00000595460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191501.hg.1 |
ENST00000596260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191501.hg.1 |
NM_000960.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000291294.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000291294.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000291294.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000291294.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000291294.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000291294.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000291294.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000594275.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000594275.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000594275.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000594275.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000595460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000595460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000595460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000595460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000595460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000595460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000595460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000596260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000596260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000596260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000596260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000596260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000597185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000597185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000597185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000597185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000597185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000598865.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000598865.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000598865.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000598865.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
ENST00000598865.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
NM_000960.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
NM_000960.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
NM_000960.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
NM_000960.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
NM_000960.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
NM_000960.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010994.hg.1 |
NM_000960.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900191505.hg.1 |
ENST00000291294.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191505.hg.1 |
ENST00000594275.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191505.hg.1 |
ENST00000596260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191505.hg.1 |
ENST00000597185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191505.hg.1 |
ENST00000598865.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191505.hg.1 |
NM_000960.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900191505.hg.1 |
TC1900010994.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900191505.hg.1 |
ENST00000291294.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191505.hg.1 |
ENST00000594275.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191505.hg.1 |
ENST00000596260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191505.hg.1 |
ENST00000597185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191505.hg.1 |
ENST00000598865.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191505.hg.1 |
NM_000960.3 |
Refseq |