Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024949.hg.1 |
TC05001828.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024949.hg.1 |
ENST00000274721.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024949.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024949.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500184327.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184327.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184327.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500184327.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184327.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184327.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500184327.hg.1 |
TC0500012164.hg.1 |
Clariom D TC |