Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013646.hg.1 |
TC06001147.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013646.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013646.hg.1 |
ENST00000472052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013646.hg.1 |
ENST00000541436.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013646.hg.1 |
NM_005891.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
ENST00000467951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
ENST00000467951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
ENST00000467951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
ENST00000467951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
ENST00000467951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
ENST00000472052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
ENST00000472052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
ENST00000472052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
ENST00000472052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
ENST00000472052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
ENST00000472052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
NM_001303253.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
NM_001303253.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
NM_001303253.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
NM_001303253.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
NM_001303253.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
NM_001303253.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
NM_001303253.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
NM_001303253.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
NM_001303253.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
NM_001303253.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
NM_001303253.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
NM_005891.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
NM_005891.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
NM_005891.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
NM_005891.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
NM_005891.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
NM_005891.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
NM_005891.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
NM_005891.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
NM_005891.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
NM_005891.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010057.hg.1 |
NM_005891.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013641.hg.1 |
TC06001147.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013641.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013641.hg.1 |
ENST00000541436.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013641.hg.1 |
NM_005891.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013642.hg.1 |
TC06001147.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013642.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013642.hg.1 |
ENST00000541436.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013642.hg.1 |
NM_005891.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168703.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600168703.hg.1 |
ENST00000467951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600168703.hg.1 |
NM_001303253.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168703.hg.1 |
NM_005891.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168703.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600168703.hg.1 |
ENST00000467951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600168703.hg.1 |
NM_001303253.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168703.hg.1 |
NM_005891.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168703.hg.1 |
TC0600010057.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600168704.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600168704.hg.1 |
ENST00000467951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600168704.hg.1 |
NM_001303253.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168704.hg.1 |
NM_005891.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168704.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600168704.hg.1 |
ENST00000467951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600168704.hg.1 |
NM_001303253.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168704.hg.1 |
NM_005891.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168704.hg.1 |
TC0600010057.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600168708.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600168708.hg.1 |
ENST00000472052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600168708.hg.1 |
NM_001303253.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168708.hg.1 |
NM_005891.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168708.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600168708.hg.1 |
ENST00000472052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600168708.hg.1 |
NM_001303253.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168708.hg.1 |
NM_005891.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168708.hg.1 |
TC0600010057.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
ENST00000367048.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
ENST00000467951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
ENST00000467951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
ENST00000467951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
ENST00000467951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
ENST00000467951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
ENST00000472052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
ENST00000472052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
ENST00000472052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
ENST00000472052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
ENST00000472052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
ENST00000472052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
NM_001303253.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
NM_001303253.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
NM_001303253.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
NM_001303253.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
NM_001303253.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
NM_001303253.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
NM_001303253.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
NM_001303253.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
NM_001303253.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
NM_001303253.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
NM_001303253.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
NM_005891.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
NM_005891.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
NM_005891.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
NM_005891.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
NM_005891.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
NM_005891.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
NM_005891.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
NM_005891.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
NM_005891.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
NM_005891.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010057.hg.1 |
NM_005891.2 |
Refseq |