Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007118.3 | NP_009049.2 | 45 - 46 | 6489 | 70 | |
NR_134469.1 | 45 - 46 | 6489 | 70 | |||
XM_011514107.2 | XP_011512409.1 | 45 - 46 | 7064 | 70 | ||
XM_011514108.1 | XP_011512410.1 | 45 - 46 | 6363 | 70 | ||
XM_011514109.2 | XP_011512411.1 | 45 - 46 | 10141 | 70 | ||
XM_011514110.2 | XP_011512412.1 | 45 - 46 | 6599 | 70 | ||
XM_017009801.1 | XP_016865290.1 | 45 - 46 | 6841 | 70 | ||
XM_017009802.1 | XP_016865291.1 | 45 - 46 | 6841 | 70 | ||
XM_017009803.1 | XP_016865292.1 | 38 - 39 | 5110 | 70 | ||
GenBank mRNA | AB209754.1 | - | 40 - 41 | 5383 | 70 | |
AF091395.1 | - | 45 - 46 | 6354 | 70 | ||
U42390.1 | - | 45 - 46 | 6354 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |