Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000355417.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000355417.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000355417.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000355417.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000355417.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000355417.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000355417.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000355417.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000355417.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000355417.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000355417.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000355417.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000355417.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000355417.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000355417.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000518189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000518189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000518189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000518189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000518189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000518189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000518189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000518189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000518189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000519448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000519448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000519448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000519448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000519448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000519740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000519740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000519740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000521308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000521308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000521308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000521308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000521308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000521308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000521308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000521308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000521308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000521308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000521308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000521308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000521308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000522179.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000522179.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000522964.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000522964.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000522964.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000522964.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000522964.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000522964.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000524344.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000524344.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000524344.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
ENST00000524344.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
NM_015621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
NM_015621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
NM_015621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
NM_015621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
NM_015621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
NM_015621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
NM_015621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
NM_015621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
NM_015621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
NM_015621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
NM_015621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
NM_015621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
NM_015621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012501.hg.1 |
NM_015621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027141.hg.1 |
TC05001949.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027141.hg.1 |
ENST00000355417.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027141.hg.1 |
ENST00000356749.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027141.hg.1 |
ENST00000521308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027141.hg.1 |
NM_015621.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500187238.hg.1 |
ENST00000355417.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500187238.hg.1 |
ENST00000518189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500187238.hg.1 |
ENST00000521308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500187238.hg.1 |
ENST00000522964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500187238.hg.1 |
NM_015621.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500187238.hg.1 |
ENST00000355417.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500187238.hg.1 |
ENST00000518189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500187238.hg.1 |
ENST00000521308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500187238.hg.1 |
ENST00000522964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500187238.hg.1 |
NM_015621.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500187238.hg.1 |
TC0500012501.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000355417.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000355417.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000355417.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000355417.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000355417.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000355417.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000355417.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000355417.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000355417.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000355417.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000355417.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000355417.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000355417.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000355417.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000355417.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000518189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000518189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000518189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000518189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000518189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000518189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000518189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000518189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000518189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000519448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000519448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000519448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000519448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000519448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000519740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000519740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000519740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000521308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000521308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000521308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000521308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000521308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000521308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000521308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000521308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000521308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000521308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000521308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000521308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000521308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000522179.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000522179.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000522964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000522964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000522964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000522964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000522964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000522964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000524344.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000524344.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000524344.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
ENST00000524344.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
NM_015621.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
NM_015621.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
NM_015621.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
NM_015621.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
NM_015621.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
NM_015621.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
NM_015621.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
NM_015621.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
NM_015621.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
NM_015621.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
NM_015621.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
NM_015621.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
NM_015621.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012501.hg.1 |
NM_015621.2 |
Refseq |