Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000454342.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000454342.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000454342.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000454342.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000595224.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000595224.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000595224.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000595224.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000595224.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000595224.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000595224.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000595224.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000595224.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000612119.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000612119.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000612119.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000612119.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000612119.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000612119.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000614074.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000614074.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000614074.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000614074.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000614074.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000614074.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000614322.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000614322.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000614322.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000614322.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000614322.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000614322.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000615626.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000615626.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000615626.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000615626.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000615626.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000615626.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000615626.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000615626.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000615915.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000615915.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000615915.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000615915.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000615915.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000615915.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000615915.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000615915.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000615915.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000615915.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000616266.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000616266.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000616266.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000616266.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000616266.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000616266.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000616266.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000616266.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000616266.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000616266.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000616266.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000616266.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000616266.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000616812.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000616812.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000616812.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000616812.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000616812.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000616812.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000616812.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000616812.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000616812.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000616812.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000616812.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000616812.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000616812.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
ENST00000616812.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
NM_001099681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
NM_001099681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
NM_001099681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
NM_001099681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
NM_001099681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
NM_001099681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
NM_001099681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
NM_001099681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
NM_001099681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
NM_001099682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
NM_001099682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
NM_001099682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
NM_001099682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
NM_001099682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
NM_001099682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
NM_001099682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
NM_001099682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
NM_001099682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
NM_024859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
NM_024859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
NM_024859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
NM_024859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
NM_024859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
NM_024859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
NM_024859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
NM_024859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
NM_024859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
NM_024859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007221.hg.1 |
NM_024859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000454342.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000454342.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000454342.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000454342.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000595224.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000595224.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000595224.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000595224.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000595224.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000595224.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000595224.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000595224.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000595224.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000612119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000612119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000612119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000612119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000612119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000612119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000614074.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000614074.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000614074.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000614074.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000614074.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000614074.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000614322.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000614322.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000614322.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000614322.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000614322.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000614322.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000615626.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000615626.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000615626.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000615626.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000615626.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000615626.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000615626.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000615626.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000615915.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000615915.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000615915.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000615915.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000615915.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000615915.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000615915.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000615915.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000615915.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000615915.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000616266.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000616266.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000616266.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000616266.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000616266.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000616266.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000616266.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000616266.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000616266.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000616266.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000616266.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000616266.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000616266.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000616812.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000616812.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000616812.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000616812.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000616812.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000616812.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000616812.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000616812.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000616812.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000616812.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000616812.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000616812.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000616812.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
ENST00000616812.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
NM_001099681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
NM_001099681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
NM_001099681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
NM_001099681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
NM_001099681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
NM_001099681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
NM_001099681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
NM_001099681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
NM_001099681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
NM_001099682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
NM_001099682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
NM_001099682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
NM_001099682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
NM_001099682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
NM_001099682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
NM_001099682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
NM_001099682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
NM_001099682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
NM_024859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
NM_024859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
NM_024859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
NM_024859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
NM_024859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
NM_024859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
NM_024859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
NM_024859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
NM_024859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
NM_024859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007221.hg.1 |
NM_024859.3 |
Refseq |