Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000466510.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000466510.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000466510.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000466510.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000479645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000479645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000479645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000479645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000479645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000479645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000491863.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000491863.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000491863.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000491863.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007625.hg.1 |
ENST00000361934.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007625.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007625.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007625.hg.1 |
TC22000394.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200154474.hg.1 |
TC2200007662.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200154474.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200154474.hg.1 |
ENST00000466510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200154474.hg.1 |
ENST00000479645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200154474.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200154474.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200154474.hg.1 |
ENST00000466510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200154474.hg.1 |
ENST00000479645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200154474.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000466510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000466510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000466510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000466510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000479645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000479645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000479645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000479645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000479645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000479645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000491863.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000491863.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000491863.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000491863.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |