Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB209277.1 | - | 21 - 22 | 3467 | 120 | |
AK295868.1 | - | 12 - 13 | 1723 | 120 | ||
BC117261.1 | - | 21 - 22 | 3403 | 40125812 | 120 | |
BC143366.1 | - | 21 - 22 | 3403 | 9051871 | 120 | |
BC143367.1 | - | 20 - 21 | 3304 | 9051872 | 120 | |
L13858.1 | - | 21 - 22 | 3377 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |