Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001184740.1 | NP_001171669.1 | 10 - 11 | 1001 | 62 | |
NM_138565.2 | NP_612632.1 | 10 - 11 | 1001 | 62 | ||
GenBank mRNA | AK291097.1 | - | 10 - 11 | 972 | 62 | |
BC008799.2 | - | 10 - 11 | 963 | 3637586 | 62 | |
BC033889.1 | - | 10 - 11 | 971 | 4824206 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |