Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008668.hg.1 |
ENST00000379589.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008668.hg.1 |
ENST00000379589.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008668.hg.1 |
ENST00000379589.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008668.hg.1 |
ENST00000379589.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008668.hg.1 |
ENST00000379589.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008668.hg.1 |
ENST00000379589.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008668.hg.1 |
ENST00000379589.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008668.hg.1 |
ENST00000379589.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008668.hg.1 |
ENST00000379589.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008668.hg.1 |
ENST00000379589.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008668.hg.1 |
ENST00000495922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008668.hg.1 |
ENST00000495922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008668.hg.1 |
ENST00000495922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008668.hg.1 |
NM_005780.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008668.hg.1 |
NM_005780.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008668.hg.1 |
NM_005780.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008668.hg.1 |
NM_005780.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008668.hg.1 |
NM_005780.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008668.hg.1 |
NM_005780.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008668.hg.1 |
NM_005780.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008668.hg.1 |
NM_005780.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008668.hg.1 |
NM_005780.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008668.hg.1 |
NM_005780.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008668.hg.1 |
NM_005780.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006728.hg.1 |
ENST00000379589.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006728.hg.1 |
NM_005780.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006728.hg.1 |
TC13000585.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300158561.hg.1 |
ENST00000379589.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158561.hg.1 |
NM_005780.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158561.hg.1 |
ENST00000379589.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158561.hg.1 |
NM_005780.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158561.hg.1 |
TC1300008668.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300008668.hg.1 |
ENST00000379589.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008668.hg.1 |
ENST00000379589.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008668.hg.1 |
ENST00000379589.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008668.hg.1 |
ENST00000379589.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008668.hg.1 |
ENST00000379589.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008668.hg.1 |
ENST00000379589.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008668.hg.1 |
ENST00000379589.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008668.hg.1 |
ENST00000379589.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008668.hg.1 |
ENST00000379589.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008668.hg.1 |
ENST00000379589.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008668.hg.1 |
ENST00000495922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008668.hg.1 |
ENST00000495922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008668.hg.1 |
ENST00000495922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008668.hg.1 |
NM_005780.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008668.hg.1 |
NM_005780.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008668.hg.1 |
NM_005780.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008668.hg.1 |
NM_005780.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008668.hg.1 |
NM_005780.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008668.hg.1 |
NM_005780.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008668.hg.1 |
NM_005780.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008668.hg.1 |
NM_005780.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008668.hg.1 |
NM_005780.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008668.hg.1 |
NM_005780.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008668.hg.1 |
NM_005780.2 |
Refseq |