Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008686.hg.1 |
TC04000598.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008686.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008686.hg.1 |
ENST00000433996.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008686.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000394488.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000394488.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000394488.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000394488.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000394488.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008687.hg.1 |
TC04000598.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008687.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008687.hg.1 |
ENST00000433996.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008687.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008684.hg.1 |
TC04000598.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008684.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008684.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400158488.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400158488.hg.1 |
ENST00000394488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400158488.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400158488.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400158488.hg.1 |
ENST00000394488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400158488.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400158488.hg.1 |
TC0400008534.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000394488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000394488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000394488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000394488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000394488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400158490.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400158490.hg.1 |
ENST00000394488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400158490.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400158490.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400158490.hg.1 |
ENST00000394488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400158490.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400158490.hg.1 |
TC0400008534.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400158487.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400158487.hg.1 |
ENST00000394488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400158487.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400158487.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400158487.hg.1 |
ENST00000394488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400158487.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400158487.hg.1 |
TC0400008534.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400158485.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400158485.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400158485.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400158485.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400158485.hg.1 |
TC0400008534.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400158486.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400158486.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400158486.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400158486.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400158486.hg.1 |
TC0400008534.hg.1 |
Clariom D TC |