Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021936.hg.1 |
TC09001719.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021936.hg.1 |
ENST00000298466.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021936.hg.1 |
ENST00000425225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021936.hg.1 |
NM_001012415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021936.hg.1 |
NM_001101677.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021937.hg.1 |
TC09001719.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021937.hg.1 |
ENST00000298466.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021937.hg.1 |
ENST00000425225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021937.hg.1 |
NM_001012415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021937.hg.1 |
NM_001101677.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900181665.hg.1 |
ENST00000298466.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181665.hg.1 |
ENST00000425225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181665.hg.1 |
NM_001012415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900181665.hg.1 |
NM_001101677.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900181665.hg.1 |
ENST00000298466.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181665.hg.1 |
ENST00000425225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181665.hg.1 |
NM_001012415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900181665.hg.1 |
NM_001101677.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900181665.hg.1 |
TC0900011950.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900181664.hg.1 |
ENST00000298466.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181664.hg.1 |
ENST00000425225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181664.hg.1 |
NM_001012415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900181664.hg.1 |
NM_001101677.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900181664.hg.1 |
ENST00000298466.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181664.hg.1 |
ENST00000425225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181664.hg.1 |
NM_001012415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900181664.hg.1 |
NM_001101677.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900181664.hg.1 |
TC0900011950.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900181666.hg.1 |
ENST00000298466.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181666.hg.1 |
ENST00000425225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181666.hg.1 |
NM_001012415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900181666.hg.1 |
NM_001101677.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900181666.hg.1 |
ENST00000298466.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181666.hg.1 |
ENST00000425225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181666.hg.1 |
NM_001012415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900181666.hg.1 |
NM_001101677.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900181666.hg.1 |
TC0900011950.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000298466.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
ENST00000425225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001012415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011950.hg.1 |
NM_001101677.1 |
Refseq |