Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008873.hg.1 |
ENST00000412060.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008873.hg.1 |
ENST00000412060.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008873.hg.1 |
ENST00000412060.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008873.hg.1 |
ENST00000412060.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008873.hg.1 |
ENST00000424852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008873.hg.1 |
ENST00000424852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008873.hg.1 |
ENST00000424852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008873.hg.1 |
ENST00000424852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008873.hg.1 |
ENST00000424852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008873.hg.1 |
ENST00000424852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008873.hg.1 |
ENST00000432473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008873.hg.1 |
ENST00000432473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008873.hg.1 |
ENST00000432473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008873.hg.1 |
ENST00000432473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008873.hg.1 |
NR_120595.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008873.hg.1 |
NR_120595.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008873.hg.1 |
NR_120595.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008873.hg.1 |
NR_120595.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008873.hg.1 |
NR_120595.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008873.hg.1 |
NR_120595.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008873.hg.1 |
NR_120596.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008873.hg.1 |
NR_120596.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008873.hg.1 |
NR_120596.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008873.hg.1 |
NR_120596.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008873.hg.1 |
NR_120596.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008873.hg.1 |
NR_120597.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008873.hg.1 |
NR_120597.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008873.hg.1 |
NR_120597.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008873.hg.1 |
NR_120597.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200164288.hg.1 |
TC2200008873.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200164288.hg.1 |
ENST00000424852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200164288.hg.1 |
ENST00000432473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200164288.hg.1 |
NR_120595.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200164288.hg.1 |
NR_120596.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200164288.hg.1 |
ENST00000424852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200164288.hg.1 |
ENST00000432473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200164288.hg.1 |
NR_120595.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200164288.hg.1 |
NR_120596.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008873.hg.1 |
ENST00000412060.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008873.hg.1 |
ENST00000412060.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008873.hg.1 |
ENST00000412060.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008873.hg.1 |
ENST00000412060.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008873.hg.1 |
ENST00000424852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008873.hg.1 |
ENST00000424852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008873.hg.1 |
ENST00000424852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008873.hg.1 |
ENST00000424852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008873.hg.1 |
ENST00000424852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008873.hg.1 |
ENST00000424852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008873.hg.1 |
ENST00000432473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008873.hg.1 |
ENST00000432473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008873.hg.1 |
ENST00000432473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008873.hg.1 |
ENST00000432473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008873.hg.1 |
NR_120595.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008873.hg.1 |
NR_120595.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008873.hg.1 |
NR_120595.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008873.hg.1 |
NR_120595.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008873.hg.1 |
NR_120595.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008873.hg.1 |
NR_120595.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008873.hg.1 |
NR_120596.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008873.hg.1 |
NR_120596.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008873.hg.1 |
NR_120596.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008873.hg.1 |
NR_120596.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008873.hg.1 |
NR_120596.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008873.hg.1 |
NR_120597.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008873.hg.1 |
NR_120597.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008873.hg.1 |
NR_120597.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008873.hg.1 |
NR_120597.1 |
Refseq |