Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010396.hg.1 |
ENST00000377024.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010396.hg.1 |
ENST00000377024.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010396.hg.1 |
ENST00000377024.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010396.hg.1 |
ENST00000377024.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010396.hg.1 |
ENST00000377024.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010396.hg.1 |
ENST00000377024.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010396.hg.1 |
ENST00000377024.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010396.hg.1 |
ENST00000424431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010396.hg.1 |
ENST00000424431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010396.hg.1 |
ENST00000424431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010396.hg.1 |
ENST00000424431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010396.hg.1 |
ENST00000424431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010396.hg.1 |
ENST00000424431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010396.hg.1 |
NM_001128618.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010396.hg.1 |
NM_001128618.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010396.hg.1 |
NM_001128618.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010396.hg.1 |
NM_001128618.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010396.hg.1 |
NM_001128618.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010396.hg.1 |
NM_001128618.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010396.hg.1 |
NM_001128618.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015473.hg.1 |
TC09001207.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015473.hg.1 |
ENST00000377024.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015473.hg.1 |
ENST00000424431.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015473.hg.1 |
NM_001128618.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015474.hg.1 |
TC09001207.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015474.hg.1 |
ENST00000377024.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015474.hg.1 |
ENST00000424431.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015474.hg.1 |
NM_001128618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900171139.hg.1 |
ENST00000377024.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171139.hg.1 |
ENST00000424431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171139.hg.1 |
NM_001128618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900171139.hg.1 |
ENST00000377024.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171139.hg.1 |
ENST00000424431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171139.hg.1 |
NM_001128618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900171139.hg.1 |
TC0900010396.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900171138.hg.1 |
ENST00000377024.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171138.hg.1 |
ENST00000424431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171138.hg.1 |
NM_001128618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900171138.hg.1 |
ENST00000377024.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171138.hg.1 |
ENST00000424431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171138.hg.1 |
NM_001128618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900171138.hg.1 |
TC0900010396.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900010396.hg.1 |
ENST00000377024.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010396.hg.1 |
ENST00000377024.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010396.hg.1 |
ENST00000377024.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010396.hg.1 |
ENST00000377024.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010396.hg.1 |
ENST00000377024.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010396.hg.1 |
ENST00000377024.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010396.hg.1 |
ENST00000377024.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010396.hg.1 |
ENST00000424431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010396.hg.1 |
ENST00000424431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010396.hg.1 |
ENST00000424431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010396.hg.1 |
ENST00000424431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010396.hg.1 |
ENST00000424431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010396.hg.1 |
ENST00000424431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010396.hg.1 |
NM_001128618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010396.hg.1 |
NM_001128618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010396.hg.1 |
NM_001128618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010396.hg.1 |
NM_001128618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010396.hg.1 |
NM_001128618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010396.hg.1 |
NM_001128618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010396.hg.1 |
NM_001128618.1 |
Refseq |