Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011705.hg.1 |
ENST00000421545.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011705.hg.1 |
ENST00000421545.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011705.hg.1 |
ENST00000421545.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011705.hg.1 |
ENST00000421545.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011705.hg.1 |
ENST00000421545.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011705.hg.1 |
ENST00000421545.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011705.hg.1 |
ENST00000421545.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011705.hg.1 |
ENST00000421545.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011705.hg.1 |
ENST00000421545.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011705.hg.1 |
ENST00000421545.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011705.hg.1 |
ENST00000421545.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011705.hg.1 |
ENST00000421545.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011705.hg.1 |
NM_001110822.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011705.hg.1 |
NM_001110822.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011705.hg.1 |
NM_001110822.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011705.hg.1 |
NM_001110822.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011705.hg.1 |
NM_001110822.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011705.hg.1 |
NM_001110822.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011705.hg.1 |
NM_001110822.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011705.hg.1 |
NM_001110822.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011705.hg.1 |
NM_001110822.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011705.hg.1 |
NM_001110822.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011705.hg.1 |
NM_001110822.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011705.hg.1 |
NM_001110822.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011705.hg.1 |
NM_001110822.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005587.hg.1 |
ENST00000421545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005587.hg.1 |
ENST00000444215.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005587.hg.1 |
NM_001110822.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005587.hg.1 |
TC19000430.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900200251.hg.1 |
ENST00000421545.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900200251.hg.1 |
NM_001110822.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900200251.hg.1 |
TC1900011705.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900200251.hg.1 |
ENST00000421545.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900200251.hg.1 |
NM_001110822.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011705.hg.1 |
ENST00000421545.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011705.hg.1 |
ENST00000421545.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011705.hg.1 |
ENST00000421545.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011705.hg.1 |
ENST00000421545.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011705.hg.1 |
ENST00000421545.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011705.hg.1 |
ENST00000421545.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011705.hg.1 |
ENST00000421545.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011705.hg.1 |
ENST00000421545.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011705.hg.1 |
ENST00000421545.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011705.hg.1 |
ENST00000421545.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011705.hg.1 |
ENST00000421545.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011705.hg.1 |
ENST00000421545.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011705.hg.1 |
NM_001110822.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011705.hg.1 |
NM_001110822.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011705.hg.1 |
NM_001110822.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011705.hg.1 |
NM_001110822.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011705.hg.1 |
NM_001110822.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011705.hg.1 |
NM_001110822.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011705.hg.1 |
NM_001110822.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011705.hg.1 |
NM_001110822.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011705.hg.1 |
NM_001110822.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011705.hg.1 |
NM_001110822.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011705.hg.1 |
NM_001110822.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011705.hg.1 |
NM_001110822.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011705.hg.1 |
NM_001110822.1 |
Refseq |