Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007961.hg.1 |
ENST00000328828.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007961.hg.1 |
ENST00000328828.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007961.hg.1 |
ENST00000328828.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007961.hg.1 |
ENST00000328828.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007961.hg.1 |
ENST00000328828.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007961.hg.1 |
ENST00000328828.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007961.hg.1 |
ENST00000328828.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007961.hg.1 |
ENST00000328828.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007961.hg.1 |
ENST00000328828.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007961.hg.1 |
ENST00000409568.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007961.hg.1 |
ENST00000409568.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007961.hg.1 |
ENST00000409568.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007961.hg.1 |
ENST00000409568.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007961.hg.1 |
ENST00000409568.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007961.hg.1 |
ENST00000409568.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007961.hg.1 |
ENST00000409568.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007961.hg.1 |
ENST00000409568.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007961.hg.1 |
NM_001101404.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007961.hg.1 |
NM_001101404.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007961.hg.1 |
NM_001101404.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007961.hg.1 |
NM_001101404.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007961.hg.1 |
NM_001101404.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007961.hg.1 |
NM_001101404.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007961.hg.1 |
NM_001101404.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15006428.hg.1 |
ENST00000328828.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15006428.hg.1 |
ENST00000409568.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15006428.hg.1 |
NM_001101404.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15006428.hg.1 |
TC15000723.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15006426.hg.1 |
ENST00000328828.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15006426.hg.1 |
NM_001101404.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15006426.hg.1 |
TC15000723.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500157059.hg.1 |
ENST00000328828.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500157059.hg.1 |
ENST00000409568.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500157059.hg.1 |
NM_001101404.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500157059.hg.1 |
ENST00000328828.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500157059.hg.1 |
ENST00000409568.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500157059.hg.1 |
NM_001101404.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500157059.hg.1 |
TC1500007961.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500157057.hg.1 |
ENST00000328828.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500157057.hg.1 |
NM_001101404.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500157057.hg.1 |
ENST00000328828.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500157057.hg.1 |
NM_001101404.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500157057.hg.1 |
TC1500007961.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500007961.hg.1 |
ENST00000328828.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007961.hg.1 |
ENST00000328828.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007961.hg.1 |
ENST00000328828.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007961.hg.1 |
ENST00000328828.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007961.hg.1 |
ENST00000328828.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007961.hg.1 |
ENST00000328828.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007961.hg.1 |
ENST00000328828.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007961.hg.1 |
ENST00000328828.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007961.hg.1 |
ENST00000328828.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007961.hg.1 |
ENST00000409568.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007961.hg.1 |
ENST00000409568.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007961.hg.1 |
ENST00000409568.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007961.hg.1 |
ENST00000409568.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007961.hg.1 |
ENST00000409568.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007961.hg.1 |
ENST00000409568.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007961.hg.1 |
ENST00000409568.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007961.hg.1 |
ENST00000409568.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007961.hg.1 |
NM_001101404.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007961.hg.1 |
NM_001101404.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007961.hg.1 |
NM_001101404.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007961.hg.1 |
NM_001101404.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007961.hg.1 |
NM_001101404.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007961.hg.1 |
NM_001101404.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007961.hg.1 |
NM_001101404.1 |
Refseq |