Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
ENST00000439350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
ENST00000439350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
ENST00000439350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
ENST00000439350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
ENST00000439350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
ENST00000439350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
ENST00000514672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
ENST00000514672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
ENST00000514672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
ENST00000514672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
ENST00000514672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
ENST00000514672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
NM_001017971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
NM_001017971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
NM_001017971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
NM_001017971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
NM_001017971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
NM_001017971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
NM_001017971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
NM_001017971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
NM_001017971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
NM_001017971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
NM_001017971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
NM_001017971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
NM_001017971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
NM_001017971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013195.hg.1 |
NM_001017971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
ENST00000439350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
ENST00000439350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
ENST00000439350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
ENST00000439350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
ENST00000439350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
ENST00000439350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
ENST00000514672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
ENST00000514672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
ENST00000514672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
ENST00000514672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
ENST00000514672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
ENST00000514672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
NM_001017971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
NM_001017971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
NM_001017971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
NM_001017971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
NM_001017971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
NM_001017971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
NM_001017971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
NM_001017971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
NM_001017971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
NM_001017971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
NM_001017971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
NM_001017971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
NM_001017971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
NM_001017971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013195.hg.1 |
NM_001017971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193413.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500193413.hg.1 |
ENST00000439350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500193413.hg.1 |
ENST00000514672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500193413.hg.1 |
NM_001017971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193413.hg.1 |
ENST00000380167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500193413.hg.1 |
ENST00000439350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500193413.hg.1 |
ENST00000514672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500193413.hg.1 |
NM_001017971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193413.hg.1 |
TC0500013195.hg.1 |
Clariom D TC |