Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
8059853 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8059852 |
- |
- |
Affymetrix Human Genome U219 Array |
11753965_PM_at |
- |
- |
Affymetrix Human Genome U219 Array |
11753965_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046466.hg.1 |
TC02002887.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046466.hg.1 |
ENST00000438684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046466.hg.1 |
NR_024322.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046468.hg.1 |
TC02002887.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046468.hg.1 |
ENST00000438684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046468.hg.1 |
NR_024322.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016100.hg.1 |
ENST00000438684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016100.hg.1 |
ENST00000438684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016100.hg.1 |
ENST00000438684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016100.hg.1 |
ENST00000438684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016100.hg.1 |
ENST00000438684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016100.hg.1 |
ENST00000440028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016100.hg.1 |
ENST00000440028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016100.hg.1 |
ENST00000440028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016100.hg.1 |
ENST00000623215.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016100.hg.1 |
ENST00000623215.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016100.hg.1 |
NR_024322.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016100.hg.1 |
NR_024322.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016100.hg.1 |
NR_024322.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016100.hg.1 |
NR_024322.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046465.hg.1 |
TC02002887.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046465.hg.1 |
ENST00000438684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046465.hg.1 |
NR_024322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200218271.hg.1 |
ENST00000438684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218271.hg.1 |
NR_024322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200218271.hg.1 |
ENST00000438684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218271.hg.1 |
NR_024322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200218271.hg.1 |
TC0200016100.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200016100.hg.1 |
ENST00000438684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016100.hg.1 |
ENST00000438684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016100.hg.1 |
ENST00000438684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016100.hg.1 |
ENST00000438684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016100.hg.1 |
ENST00000438684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016100.hg.1 |
ENST00000440028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016100.hg.1 |
ENST00000440028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016100.hg.1 |
ENST00000440028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016100.hg.1 |
ENST00000623215.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016100.hg.1 |
ENST00000623215.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016100.hg.1 |
NR_024322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016100.hg.1 |
NR_024322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016100.hg.1 |
NR_024322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016100.hg.1 |
NR_024322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200218265.hg.1 |
ENST00000438684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218265.hg.1 |
NR_024322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200218265.hg.1 |
ENST00000438684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218265.hg.1 |
NR_024322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200218265.hg.1 |
TC0200016100.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200218267.hg.1 |
ENST00000438684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218267.hg.1 |
ENST00000440028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218267.hg.1 |
ENST00000623215.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218267.hg.1 |
NR_024322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200218267.hg.1 |
ENST00000438684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218267.hg.1 |
ENST00000440028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218267.hg.1 |
ENST00000623215.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218267.hg.1 |
NR_024322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200218267.hg.1 |
TC0200016100.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200218266.hg.1 |
ENST00000438684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218266.hg.1 |
ENST00000623215.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218266.hg.1 |
NR_024322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200218266.hg.1 |
ENST00000438684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218266.hg.1 |
ENST00000623215.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218266.hg.1 |
NR_024322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200218266.hg.1 |
TC0200016100.hg.1 |
Clariom D TC |