Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15193235 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15193234 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20943227 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872763 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872720 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872756 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20943235 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20943248 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872743 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15193227 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20943260 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872744 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872758 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15193226 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872757 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20943291 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872704 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872716 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872732 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872714 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20943241 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20943293 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872766 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15193232 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20943258 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872715 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20943283 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872734 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20943257 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872708 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20943249 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20943261 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872747 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872722 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20943282 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872749 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20943277 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872700 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20943274 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872761 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20943246 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872723 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872751 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872737 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872710 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20943259 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872703 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872694 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20943239 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20943244 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872724 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872768 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872717 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20943289 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20943267 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872741 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872709 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872695 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20943240 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872697 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20943221 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872752 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20943229 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00872733 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
159672 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 8x60K |
48821 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
291547 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
454448 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
365256 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
163983 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
108326 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
16018 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
164250 |
AK056964.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
164250 |
AK313410.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
164250 |
BC002328.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
164250 |
BC010184.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
164250 |
BC075810.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
164250 |
NM_022758.4 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
164250 |
NM_024294.2 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
227024 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
420049 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
103798 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
356371 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
339276 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P63459 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_23_P63459 |
- |
- |
Agilent Whole Human Genome Oligo Microarray 4x44K |
4529 |
- |
- |
Agilent Human Genome CGH Microarray 44B |
A_14_P126554 |
- |
- |
Agilent Human Genome CGH Microarray 44A |
A_14_P126554 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
26007 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_18_P10555009 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_18_P18134703 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P30497513 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P00277670 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P63459 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_23_P63459 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943248 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943261 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943257 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943230 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943259 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00872722 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00872709 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00872714 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00872708 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00872733 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00872710 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15193235 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943245 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943224 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00872717 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943241 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943244 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943246 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943227 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00872695 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00872724 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943258 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15193227 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943260 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15193228 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943229 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00872697 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00872723 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00872706 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00872704 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943239 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00872703 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15193234 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943249 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00872700 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943250 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15193232 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00872732 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943249 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00872732 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00872697 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15193235 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00872722 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00872708 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15193228 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943250 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943241 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00872714 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00872723 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943261 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943257 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943260 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943248 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943259 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943239 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943245 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943229 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00872709 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00872717 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00872733 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943230 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943246 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00872704 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15193227 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943244 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00872710 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00872695 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00872703 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00872700 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943258 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943227 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00872724 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00872706 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15193232 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15193234 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20943224 |
- |
- |
Whole Human Genome Oligo Microarray G4112A |
A_23_P63459 |
- |
- |
Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P126554 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00872708 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00872722 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00872716 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00872718 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00872724 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00872727 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00872706 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00872720 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00872702 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00872730 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00872704 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00872731 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00872713 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00872703 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00872711 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00872721 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00872699 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00872725 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00872733 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15193233 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P00872722 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P00872724 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15193232 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15193234 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P00872723 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P00277670 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P126554 |
- |
- |
Agilent Human Genome CGH Microarray 4x44K |
A_14_P126554 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K G4112F |
A_23_P63459 |
- |
- |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
28546 |
- |
- |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
3773 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P15490164 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_14_P126554 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P18134016 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P00277670 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872718 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872708 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872702 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872749 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872733 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872693 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872742 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872716 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872710 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872722 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872727 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872766 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872720 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872737 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872731 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872751 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872730 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872706 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872709 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872740 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872699 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872745 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872695 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872756 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872713 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872735 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872724 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15193226 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15193232 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872768 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15193231 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872761 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872725 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872758 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872711 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872704 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872739 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872747 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872697 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15193234 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00872753 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_14_P126554 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P00277670 |
- |
- |