Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPEG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005876.4 | NP_005867.3 | 35 - 36 | 8323 | 82 | |
XM_005246237.2 | XP_005246294.1 | 37 - 38 | 8051 | 82 | ||
XM_005246239.2 | XP_005246296.1 | 37 - 38 | 7866 | 82 | ||
XM_005246240.2 | XP_005246297.1 | 30 - 31 | 6068 | 82 | ||
XM_005246241.1 | XP_005246298.1 | 30 - 31 | 5962 | 82 | ||
XM_005246242.4 | XP_005246299.1 | 30 - 31 | 6087 | 82 | ||
XM_006712189.3 | XP_006712252.1 | 36 - 37 | 8056 | 82 | ||
XM_006712193.3 | XP_006712256.1 | 30 - 31 | 9187 | 82 | ||
XM_011510479.2 | XP_011508781.1 | 36 - 37 | 8489 | 82 | ||
XM_011510483.2 | XP_011508785.2 | 36 - 37 | 8086 | 82 | ||
XM_017003157.1 | XP_016858646.1 | 36 - 37 | 8086 | 82 | ||
XM_017003158.1 | XP_016858647.1 | 32 - 33 | 7662 | 82 | ||
XM_017003160.1 | XP_016858649.1 | 20 - 21 | 4258 | 82 | ||
GenBank mRNA | AB037718.1 | - | 26 - 27 | 5380 | 82 | |
AY603755.1 | - | 34 - 35 | 7936 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Homo sapiens Probe spans exons |