Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006782.hg.1 |
ENST00000385060.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006782.hg.1 |
ENST00000456917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006782.hg.1 |
ENST00000456917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006782.hg.1 |
ENST00000456917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006782.hg.1 |
ENST00000456917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006782.hg.1 |
ENST00000456917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006782.hg.1 |
ENST00000456917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006782.hg.1 |
NR_001458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006782.hg.1 |
NR_001458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006782.hg.1 |
NR_001458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006782.hg.1 |
NR_001458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006782.hg.1 |
NR_001458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006782.hg.1 |
NR_030784.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000380.hg.1 |
ENST00000456917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000380.hg.1 |
NR_001458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000380.hg.1 |
TC21000076.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000379.hg.1 |
ENST00000456917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000379.hg.1 |
NR_001458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000379.hg.1 |
TC21000076.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2100145740.hg.1 |
ENST00000456917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100145740.hg.1 |
NR_001458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100145740.hg.1 |
TC2100006782.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100145740.hg.1 |
ENST00000456917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100145740.hg.1 |
NR_001458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100145742.hg.1 |
ENST00000456917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100145742.hg.1 |
NR_001458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100145742.hg.1 |
TC2100006782.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100145742.hg.1 |
ENST00000456917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100145742.hg.1 |
NR_001458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006782.hg.1 |
ENST00000385060.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006782.hg.1 |
ENST00000456917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006782.hg.1 |
ENST00000456917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006782.hg.1 |
ENST00000456917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006782.hg.1 |
ENST00000456917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006782.hg.1 |
ENST00000456917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006782.hg.1 |
ENST00000456917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006782.hg.1 |
NR_001458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006782.hg.1 |
NR_001458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006782.hg.1 |
NR_001458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006782.hg.1 |
NR_001458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006782.hg.1 |
NR_001458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006782.hg.1 |
NR_030784.1 |
Refseq |