Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019292.hg.1 |
NM_001166692.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019292.hg.1 |
TC11001540.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019294.hg.1 |
ENST00000379011.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019294.hg.1 |
ENST00000527926.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019294.hg.1 |
ENST00000534312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019294.hg.1 |
NM_001166692.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019294.hg.1 |
TC11001540.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013160.hg.1 |
ENST00000379011.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013160.hg.1 |
ENST00000379011.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013160.hg.1 |
ENST00000379011.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013160.hg.1 |
ENST00000379011.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013160.hg.1 |
ENST00000379011.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013160.hg.1 |
ENST00000379011.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013160.hg.1 |
ENST00000531145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013160.hg.1 |
ENST00000531145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013160.hg.1 |
ENST00000531145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013160.hg.1 |
NM_001166692.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013160.hg.1 |
NM_001166692.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013160.hg.1 |
NM_001166692.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013160.hg.1 |
NM_001166692.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013160.hg.1 |
NM_001166692.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013160.hg.1 |
NM_001166692.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019297.hg.1 |
ENST00000379011.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019297.hg.1 |
ENST00000451594.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019297.hg.1 |
NM_001166692.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019297.hg.1 |
TC11001540.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1100013160.hg.1 |
ENST00000379011.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013160.hg.1 |
ENST00000379011.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013160.hg.1 |
ENST00000379011.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013160.hg.1 |
ENST00000379011.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013160.hg.1 |
ENST00000379011.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013160.hg.1 |
ENST00000379011.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013160.hg.1 |
ENST00000531145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013160.hg.1 |
ENST00000531145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013160.hg.1 |
ENST00000531145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013160.hg.1 |
NM_001166692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013160.hg.1 |
NM_001166692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013160.hg.1 |
NM_001166692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013160.hg.1 |
NM_001166692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013160.hg.1 |
NM_001166692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013160.hg.1 |
NM_001166692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203979.hg.1 |
ENST00000379011.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203979.hg.1 |
NM_001166692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203979.hg.1 |
ENST00000379011.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203979.hg.1 |
NM_001166692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203979.hg.1 |
TC1100013160.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100203980.hg.1 |
ENST00000379011.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203980.hg.1 |
NM_001166692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203980.hg.1 |
ENST00000379011.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203980.hg.1 |
NM_001166692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203980.hg.1 |
TC1100013160.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100203986.hg.1 |
ENST00000379011.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203986.hg.1 |
ENST00000531145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203986.hg.1 |
NM_001166692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203986.hg.1 |
ENST00000379011.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203986.hg.1 |
ENST00000531145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203986.hg.1 |
NM_001166692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203986.hg.1 |
TC1100013160.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100203978.hg.1 |
NM_001166692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203978.hg.1 |
NM_001166692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203978.hg.1 |
TC1100013160.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100203982.hg.1 |
ENST00000379011.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203982.hg.1 |
ENST00000531145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203982.hg.1 |
NM_001166692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203982.hg.1 |
ENST00000379011.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203982.hg.1 |
ENST00000531145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203982.hg.1 |
NM_001166692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203982.hg.1 |
TC1100013160.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100203988.hg.1 |
ENST00000379011.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203988.hg.1 |
NM_001166692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203988.hg.1 |
ENST00000379011.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203988.hg.1 |
NM_001166692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203988.hg.1 |
TC1100013160.hg.1 |
Clariom D TC |