Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010295.hg.1 |
ENST00000532874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010295.hg.1 |
ENST00000532874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010295.hg.1 |
ENST00000532874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010295.hg.1 |
NR_033989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010295.hg.1 |
NR_033989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010295.hg.1 |
NR_033989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010295.hg.1 |
NR_033989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11018743.hg.1 |
NR_033989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11018743.hg.1 |
TC11001481.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11018741.hg.1 |
ENST00000532874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11018741.hg.1 |
NR_033989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11018741.hg.1 |
TC11001481.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1100010295.hg.1 |
ENST00000532874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010295.hg.1 |
ENST00000532874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010295.hg.1 |
ENST00000532874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010295.hg.1 |
NR_033989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010295.hg.1 |
NR_033989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010295.hg.1 |
NR_033989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010295.hg.1 |
NR_033989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100178348.hg.1 |
ENST00000532874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100178348.hg.1 |
NR_033989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100178348.hg.1 |
ENST00000532874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100178348.hg.1 |
NR_033989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100178348.hg.1 |
TC1100010295.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100178350.hg.1 |
NR_033989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100178350.hg.1 |
NR_033989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100178350.hg.1 |
TC1100010295.hg.1 |
Clariom D TC |