Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001285.hg.1 |
ENST00000262487.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001285.hg.1 |
NM_080826.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001285.hg.1 |
TC20000094.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006743.hg.1 |
ENST00000262487.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006743.hg.1 |
ENST00000262487.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006743.hg.1 |
ENST00000262487.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006743.hg.1 |
ENST00000262487.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006743.hg.1 |
ENST00000262487.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006743.hg.1 |
ENST00000262487.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006743.hg.1 |
ENST00000262487.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006743.hg.1 |
NM_080826.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006743.hg.1 |
NM_080826.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006743.hg.1 |
NM_080826.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006743.hg.1 |
NM_080826.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006743.hg.1 |
NM_080826.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006743.hg.1 |
NM_080826.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006743.hg.1 |
NM_080826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000146240.hg.1 |
ENST00000262487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146240.hg.1 |
NM_080826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000146240.hg.1 |
TC2000006743.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000146240.hg.1 |
ENST00000262487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146240.hg.1 |
NM_080826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006743.hg.1 |
ENST00000262487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006743.hg.1 |
ENST00000262487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006743.hg.1 |
ENST00000262487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006743.hg.1 |
ENST00000262487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006743.hg.1 |
ENST00000262487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006743.hg.1 |
ENST00000262487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006743.hg.1 |
ENST00000262487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006743.hg.1 |
NM_080826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006743.hg.1 |
NM_080826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006743.hg.1 |
NM_080826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006743.hg.1 |
NM_080826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006743.hg.1 |
NM_080826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006743.hg.1 |
NM_080826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006743.hg.1 |
NM_080826.1 |
Refseq |