Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007256.hg.1 |
ENST00000456123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007256.hg.1 |
ENST00000456123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007256.hg.1 |
ENST00000456123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007256.hg.1 |
ENST00000456123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006805.hg.1 |
ENST00000420149.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006805.hg.1 |
ENST00000420149.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006805.hg.1 |
ENST00000420149.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006805.hg.1 |
ENST00000420149.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001038.hg.1 |
TC0Y000091.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001038.hg.1 |
ENST00000420149.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001038.hg.1 |
NR_001525.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001038.hg.1 |
NR_002177.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001038.hg.1 |
NR_002178.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006773.hg.1 |
ENST00000436568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006773.hg.1 |
ENST00000436568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006773.hg.1 |
ENST00000436568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006773.hg.1 |
ENST00000436568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006773.hg.1 |
NR_001525.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006773.hg.1 |
NR_001525.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006773.hg.1 |
NR_001525.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006773.hg.1 |
NR_001525.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006773.hg.1 |
NR_002177.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006773.hg.1 |
NR_002177.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006773.hg.1 |
NR_002177.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006773.hg.1 |
NR_002177.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006773.hg.1 |
NR_002178.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006773.hg.1 |
NR_002178.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006773.hg.1 |
NR_002178.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006773.hg.1 |
NR_002178.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001039.hg.1 |
TC0Y000091.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001039.hg.1 |
ENST00000420149.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001039.hg.1 |
NR_001525.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001039.hg.1 |
NR_002177.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001039.hg.1 |
NR_002178.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00146129.hg.1 |
ENST00000436568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00146129.hg.1 |
NR_001525.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00146129.hg.1 |
NR_002177.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00146129.hg.1 |
NR_002178.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00146129.hg.1 |
ENST00000436568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00146129.hg.1 |
NR_001525.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00146129.hg.1 |
NR_002177.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00146129.hg.1 |
NR_002178.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00146129.hg.1 |
TC0Y00006773.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0Y00007256.hg.1 |
ENST00000456123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007256.hg.1 |
ENST00000456123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007256.hg.1 |
ENST00000456123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007256.hg.1 |
ENST00000456123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00146128.hg.1 |
ENST00000436568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00146128.hg.1 |
NR_001525.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00146128.hg.1 |
NR_002177.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00146128.hg.1 |
NR_002178.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00146128.hg.1 |
ENST00000436568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00146128.hg.1 |
NR_001525.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00146128.hg.1 |
NR_002177.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00146128.hg.1 |
NR_002178.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00146128.hg.1 |
TC0Y00006773.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0Y00006773.hg.1 |
ENST00000436568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006773.hg.1 |
ENST00000436568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006773.hg.1 |
ENST00000436568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006773.hg.1 |
ENST00000436568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006773.hg.1 |
NR_001525.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006773.hg.1 |
NR_001525.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006773.hg.1 |
NR_001525.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006773.hg.1 |
NR_001525.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006773.hg.1 |
NR_002177.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006773.hg.1 |
NR_002177.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006773.hg.1 |
NR_002177.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006773.hg.1 |
NR_002177.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006773.hg.1 |
NR_002178.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006773.hg.1 |
NR_002178.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006773.hg.1 |
NR_002178.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006773.hg.1 |
NR_002178.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006805.hg.1 |
ENST00000420149.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006805.hg.1 |
ENST00000420149.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006805.hg.1 |
ENST00000420149.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006805.hg.1 |
ENST00000420149.1 |
Ensembl |