Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
ENST00000279873.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
ENST00000279873.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
ENST00000279873.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
ENST00000279873.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
ENST00000279873.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
ENST00000279873.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
ENST00000279873.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
ENST00000279873.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
ENST00000279873.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
ENST00000279873.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
ENST00000279873.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
ENST00000279873.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
ENST00000309334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
ENST00000309334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
ENST00000309334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
ENST00000309334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
ENST00000309334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
ENST00000309334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
ENST00000309334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
ENST00000309334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
NM_001244638.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
NM_001244638.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
NM_001244638.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
NM_001244638.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
NM_001244638.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
NM_001244638.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
NM_001244638.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
NM_001244638.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
NM_001244638.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
NM_032199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
NM_032199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
NM_032199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
NM_032199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
NM_032199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
NM_032199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
NM_032199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
NM_032199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
NM_032199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
NM_032199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
NM_032199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
NM_032199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007761.hg.1 |
NM_032199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004300.hg.1 |
ENST00000279873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004300.hg.1 |
ENST00000309334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004300.hg.1 |
NM_001244638.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004300.hg.1 |
NM_032199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004300.hg.1 |
TC10000377.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000152643.hg.1 |
ENST00000279873.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152643.hg.1 |
ENST00000309334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152643.hg.1 |
NM_001244638.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152643.hg.1 |
NM_032199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152643.hg.1 |
ENST00000279873.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152643.hg.1 |
ENST00000309334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152643.hg.1 |
NM_001244638.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152643.hg.1 |
NM_032199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152643.hg.1 |
TC1000007761.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
ENST00000279873.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
ENST00000279873.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
ENST00000279873.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
ENST00000279873.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
ENST00000279873.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
ENST00000279873.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
ENST00000279873.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
ENST00000279873.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
ENST00000279873.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
ENST00000279873.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
ENST00000279873.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
ENST00000279873.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
ENST00000309334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
ENST00000309334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
ENST00000309334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
ENST00000309334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
ENST00000309334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
ENST00000309334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
ENST00000309334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
ENST00000309334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
NM_001244638.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
NM_001244638.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
NM_001244638.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
NM_001244638.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
NM_001244638.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
NM_001244638.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
NM_001244638.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
NM_001244638.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
NM_001244638.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
NM_032199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
NM_032199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
NM_032199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
NM_032199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
NM_032199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
NM_032199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
NM_032199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
NM_032199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
NM_032199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
NM_032199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
NM_032199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
NM_032199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007761.hg.1 |
NM_032199.2 |
Refseq |