Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009554.hg.1 |
TC06000796.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009554.hg.1 |
ENST00000275072.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009554.hg.1 |
NM_001010853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008760.hg.1 |
ENST00000275072.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008760.hg.1 |
ENST00000275072.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008760.hg.1 |
ENST00000275072.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008760.hg.1 |
ENST00000275072.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008760.hg.1 |
ENST00000275072.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008760.hg.1 |
ENST00000275072.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008760.hg.1 |
ENST00000275072.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008760.hg.1 |
ENST00000275072.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008760.hg.1 |
ENST00000275072.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008760.hg.1 |
NM_001010853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008760.hg.1 |
NM_001010853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008760.hg.1 |
NM_001010853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008760.hg.1 |
NM_001010853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008760.hg.1 |
NM_001010853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008760.hg.1 |
NM_001010853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008760.hg.1 |
NM_001010853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008760.hg.1 |
NM_001010853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008760.hg.1 |
NM_001010853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009555.hg.1 |
TC06000796.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009555.hg.1 |
ENST00000275072.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009555.hg.1 |
NM_001010853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600160805.hg.1 |
ENST00000275072.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600160805.hg.1 |
NM_001010853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600160805.hg.1 |
ENST00000275072.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600160805.hg.1 |
NM_001010853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600160805.hg.1 |
TC0600008760.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600160806.hg.1 |
ENST00000275072.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600160806.hg.1 |
NM_001010853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600160806.hg.1 |
ENST00000275072.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600160806.hg.1 |
NM_001010853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600160806.hg.1 |
TC0600008760.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600160804.hg.1 |
ENST00000275072.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600160804.hg.1 |
NM_001010853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600160804.hg.1 |
ENST00000275072.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600160804.hg.1 |
NM_001010853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600160804.hg.1 |
TC0600008760.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600008760.hg.1 |
ENST00000275072.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008760.hg.1 |
ENST00000275072.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008760.hg.1 |
ENST00000275072.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008760.hg.1 |
ENST00000275072.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008760.hg.1 |
ENST00000275072.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008760.hg.1 |
ENST00000275072.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008760.hg.1 |
ENST00000275072.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008760.hg.1 |
ENST00000275072.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008760.hg.1 |
ENST00000275072.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008760.hg.1 |
NM_001010853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008760.hg.1 |
NM_001010853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008760.hg.1 |
NM_001010853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008760.hg.1 |
NM_001010853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008760.hg.1 |
NM_001010853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008760.hg.1 |
NM_001010853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008760.hg.1 |
NM_001010853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008760.hg.1 |
NM_001010853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008760.hg.1 |
NM_001010853.2 |
Refseq |