Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006536.hg.1 |
ENST00000585643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006536.hg.1 |
ENST00000585643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008548.hg.1 |
ENST00000428419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008548.hg.1 |
ENST00000428419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008548.hg.1 |
ENST00000428419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008548.hg.1 |
ENST00000428419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008548.hg.1 |
ENST00000428419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008548.hg.1 |
NR_103839.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008548.hg.1 |
NR_103839.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008548.hg.1 |
NR_103839.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008548.hg.1 |
NR_103839.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008548.hg.1 |
NR_103839.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008548.hg.1 |
NR_103839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006536.hg.1 |
ENST00000585643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006536.hg.1 |
ENST00000585643.1 |
Ensembl |