Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000536603.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000536603.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000536603.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000536603.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000536603.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000536603.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000536603.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000536603.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000536603.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000536603.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000536603.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000536603.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000594087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000594087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000594087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000594087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000594087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000594087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000594087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000594087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000594087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000594672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000594672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000594672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000594672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000596161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000596161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000596161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000596161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000596687.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000596687.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000596687.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000596687.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000596687.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000596687.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000599044.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000599044.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000599044.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000599044.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000599044.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000599044.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000601763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
ENST00000601763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900164141.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164141.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900164141.hg.1 |
TC1900008358.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900164141.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164141.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000536603.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000536603.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000536603.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000536603.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000536603.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000536603.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000536603.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000536603.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000536603.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000536603.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000536603.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000536603.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000594087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000594087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000594087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000594087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000594087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000594087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000594087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000594087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000594087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000594672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000594672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000594672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000594672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000596161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000596161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000596161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000596161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000596687.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000596687.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000596687.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000596687.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000596687.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000596687.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000599044.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000599044.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000599044.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000599044.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000599044.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000599044.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000601763.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
ENST00000601763.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008358.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900164140.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164140.hg.1 |
NM_001267723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900164140.hg.1 |
TC1900008358.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900164140.hg.1 |
ENST00000595358.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164140.hg.1 |
NM_001267723.1 |
Refseq |