Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000477796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000477796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000477796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000477796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000477796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000477796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000477796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
ENST00000477796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000477796.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000477796.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000477796.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000477796.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000477796.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000477796.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000477796.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
ENST00000477796.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008070.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155179.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155179.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155179.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155179.hg.1 |
ENST00000276185.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155179.hg.1 |
ENST00000439554.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155179.hg.1 |
NM_032428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155179.hg.1 |
TC0X00008070.hg.1 |
Clariom D TC |