Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011539728.2 | XP_011538030.1 | 17 | 3744 | 165 | |
XM_017016159.1 | XP_016871648.1 | 16 | 3676 | 165 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |