Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013737.hg.1 |
TC07000881.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013737.hg.1 |
ENST00000275874.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013737.hg.1 |
ENST00000356407.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013737.hg.1 |
ENST00000537763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013737.hg.1 |
NM_001008749.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013735.hg.1 |
TC07000881.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013735.hg.1 |
ENST00000275874.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013735.hg.1 |
ENST00000537763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013735.hg.1 |
NM_001008749.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013740.hg.1 |
TC07000881.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013740.hg.1 |
ENST00000275874.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013740.hg.1 |
ENST00000356407.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013740.hg.1 |
ENST00000537763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013740.hg.1 |
NM_001008749.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009369.hg.1 |
ENST00000356407.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009369.hg.1 |
ENST00000356407.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009369.hg.1 |
ENST00000356407.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009369.hg.1 |
ENST00000356407.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009369.hg.1 |
ENST00000356407.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009369.hg.1 |
ENST00000356407.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009369.hg.1 |
ENST00000356407.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009369.hg.1 |
ENST00000495590.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009369.hg.1 |
ENST00000495590.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009369.hg.1 |
ENST00000495590.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009369.hg.1 |
ENST00000495590.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009369.hg.1 |
ENST00000495590.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009369.hg.1 |
ENST00000537763.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009369.hg.1 |
ENST00000537763.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009369.hg.1 |
ENST00000537763.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009369.hg.1 |
ENST00000537763.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009369.hg.1 |
ENST00000537763.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009369.hg.1 |
ENST00000537763.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009369.hg.1 |
ENST00000537763.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009369.hg.1 |
NM_001008749.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009369.hg.1 |
NM_001008749.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009369.hg.1 |
NM_001008749.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009369.hg.1 |
NM_001008749.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009369.hg.1 |
NM_001008749.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009369.hg.1 |
NM_001008749.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009369.hg.1 |
NM_001008749.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700166475.hg.1 |
ENST00000356407.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166475.hg.1 |
ENST00000495590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166475.hg.1 |
ENST00000537763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166475.hg.1 |
NM_001008749.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700166475.hg.1 |
ENST00000356407.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166475.hg.1 |
ENST00000495590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166475.hg.1 |
ENST00000537763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166475.hg.1 |
NM_001008749.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700166475.hg.1 |
TC0700009369.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700166477.hg.1 |
ENST00000356407.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166477.hg.1 |
ENST00000495590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166477.hg.1 |
ENST00000537763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166477.hg.1 |
NM_001008749.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700166477.hg.1 |
ENST00000356407.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166477.hg.1 |
ENST00000495590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166477.hg.1 |
ENST00000537763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166477.hg.1 |
NM_001008749.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700166477.hg.1 |
TC0700009369.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700166470.hg.1 |
ENST00000495590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166470.hg.1 |
ENST00000537763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166470.hg.1 |
NM_001008749.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700166470.hg.1 |
ENST00000495590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166470.hg.1 |
ENST00000537763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166470.hg.1 |
NM_001008749.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700166470.hg.1 |
TC0700009369.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700166471.hg.1 |
ENST00000356407.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166471.hg.1 |
ENST00000356407.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166471.hg.1 |
TC0700009369.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700166473.hg.1 |
ENST00000356407.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166473.hg.1 |
ENST00000495590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166473.hg.1 |
ENST00000537763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166473.hg.1 |
NM_001008749.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700166473.hg.1 |
ENST00000356407.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166473.hg.1 |
ENST00000495590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166473.hg.1 |
ENST00000537763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166473.hg.1 |
NM_001008749.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700166473.hg.1 |
TC0700009369.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700009369.hg.1 |
ENST00000356407.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009369.hg.1 |
ENST00000356407.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009369.hg.1 |
ENST00000356407.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009369.hg.1 |
ENST00000356407.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009369.hg.1 |
ENST00000356407.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009369.hg.1 |
ENST00000356407.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009369.hg.1 |
ENST00000356407.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009369.hg.1 |
ENST00000495590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009369.hg.1 |
ENST00000495590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009369.hg.1 |
ENST00000495590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009369.hg.1 |
ENST00000495590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009369.hg.1 |
ENST00000495590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009369.hg.1 |
ENST00000537763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009369.hg.1 |
ENST00000537763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009369.hg.1 |
ENST00000537763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009369.hg.1 |
ENST00000537763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009369.hg.1 |
ENST00000537763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009369.hg.1 |
ENST00000537763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009369.hg.1 |
ENST00000537763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009369.hg.1 |
NM_001008749.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009369.hg.1 |
NM_001008749.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009369.hg.1 |
NM_001008749.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009369.hg.1 |
NM_001008749.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009369.hg.1 |
NM_001008749.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009369.hg.1 |
NM_001008749.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009369.hg.1 |
NM_001008749.2 |
Refseq |