Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT HG-U133+ PM Array Plate |
224870_PM_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004264.hg.1 |
TC04000297.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004264.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004264.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004267.hg.1 |
TC04000297.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004267.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004267.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC04000297.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC04000297.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC04000297.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC04000297.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC04000297.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC04000297.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC04000297.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC04000297.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC04000297.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC04000297.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
ENST00000425653.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
ENST00000425653.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
ENST00000425653.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
ENST00000425653.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
ENST00000425653.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
ENST00000425653.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
ENST00000441504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
ENST00000441504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
ENST00000441504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
ENST00000444958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
ENST00000444958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
ENST00000444958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
ENST00000444958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
ENST00000578365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
ENST00000623036.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
ENST00000623036.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
ENST00000623036.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
ENST00000623036.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007511.hg.1 |
NR_039651.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004270.hg.1 |
TC04000297.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004270.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004270.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004265.hg.1 |
TC04000297.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004265.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004265.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004269.hg.1 |
TC04000297.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004269.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004269.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151329.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151329.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151329.hg.1 |
ENST00000425653.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151329.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151329.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151329.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151329.hg.1 |
ENST00000425653.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151329.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151329.hg.1 |
TC0400007511.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
ENST00000425653.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
ENST00000425653.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
ENST00000425653.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
ENST00000425653.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
ENST00000425653.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
ENST00000425653.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
ENST00000441504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
ENST00000441504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
ENST00000441504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
ENST00000444958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
ENST00000444958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
ENST00000444958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
ENST00000444958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
ENST00000578365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
ENST00000623036.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
ENST00000623036.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
ENST00000623036.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
ENST00000623036.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007511.hg.1 |
NR_039651.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151330.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151330.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151330.hg.1 |
ENST00000425653.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151330.hg.1 |
ENST00000623036.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151330.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151330.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151330.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151330.hg.1 |
ENST00000425653.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151330.hg.1 |
ENST00000623036.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151330.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151330.hg.1 |
TC0400007511.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400151315.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151315.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151315.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151315.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151315.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151315.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151315.hg.1 |
TC0400007511.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400151331.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151331.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151331.hg.1 |
ENST00000425653.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151331.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151331.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151331.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151331.hg.1 |
ENST00000425653.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151331.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151331.hg.1 |
TC0400007511.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400151316.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151316.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151316.hg.1 |
ENST00000441504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151316.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151316.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151316.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151316.hg.1 |
ENST00000441504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151316.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151316.hg.1 |
TC0400007511.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400151323.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151323.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151323.hg.1 |
ENST00000425653.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151323.hg.1 |
ENST00000441504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151323.hg.1 |
ENST00000444958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151323.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151323.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151323.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151323.hg.1 |
ENST00000425653.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151323.hg.1 |
ENST00000441504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151323.hg.1 |
ENST00000444958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151323.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151323.hg.1 |
TC0400007511.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400151328.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151328.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151328.hg.1 |
ENST00000425653.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151328.hg.1 |
ENST00000444958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151328.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151328.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151328.hg.1 |
ENST00000411630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151328.hg.1 |
ENST00000425653.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151328.hg.1 |
ENST00000444958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151328.hg.1 |
NR_024031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151328.hg.1 |
TC0400007511.hg.1 |
Clariom D TC |
Human Genome U133 Plus 2.0 Array |
224870_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
224870_PM_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224870_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224870_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224870_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224870_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224870_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224870_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224870_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224870_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224870_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224870_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224870_at9 |
- |
- |
Human Genome U95 Set |
65727_PM_at |
- |
- |
Human Genome U95 Set |
65727_at |
- |
- |
Human Genome U133 Set |
224870_PM_at |
- |
- |
Human Genome U133 Set |
224870_at |
- |
- |