Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000507440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000507440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000507440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000507440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000507440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000507440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000507440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000507440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000507440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000507440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000507440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000507440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000507440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000507440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000509536.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000509536.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000509536.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000509536.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000509536.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000511356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000511356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000511356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000511356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000511356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000511356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000511356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000511356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000511356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000511356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000511997.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000511997.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000513125.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000513125.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000513125.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000513125.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
ENST00000513125.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
NM_001010874.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
NM_001010874.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
NM_001010874.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
NM_001010874.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
NM_001010874.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
NM_001010874.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
NM_001010874.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
NM_001010874.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
NM_001010874.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
NM_001010874.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
NM_001010874.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
NM_001010874.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
NM_001010874.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
NM_001010874.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010853.hg.1 |
NM_001010874.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016868.hg.1 |
TC04001242.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016868.hg.1 |
ENST00000381210.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016868.hg.1 |
ENST00000507440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016868.hg.1 |
NM_001010874.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400173742.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400173742.hg.1 |
ENST00000507440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400173742.hg.1 |
ENST00000509536.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400173742.hg.1 |
ENST00000511356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400173742.hg.1 |
NM_001010874.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400173742.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400173742.hg.1 |
ENST00000507440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400173742.hg.1 |
ENST00000509536.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400173742.hg.1 |
ENST00000511356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400173742.hg.1 |
NM_001010874.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400173742.hg.1 |
TC0400010853.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000381210.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000507440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000507440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000507440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000507440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000507440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000507440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000507440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000507440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000507440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000507440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000507440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000507440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000507440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000507440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000509536.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000509536.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000509536.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000509536.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000509536.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000511356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000511356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000511356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000511356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000511356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000511356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000511356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000511356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000511356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000511356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000511997.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000511997.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000513125.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000513125.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000513125.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000513125.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
ENST00000513125.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
NM_001010874.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
NM_001010874.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
NM_001010874.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
NM_001010874.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
NM_001010874.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
NM_001010874.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
NM_001010874.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
NM_001010874.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
NM_001010874.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
NM_001010874.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
NM_001010874.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
NM_001010874.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
NM_001010874.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
NM_001010874.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010853.hg.1 |
NM_001010874.4 |
Refseq |