Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIPK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144685.4 | NP_653286.2 | 2 - 3 | 1144 | 95 | |
XM_006723036.2 | XP_006723099.1 | 2 - 3 | 1119 | 95 | ||
GenBank mRNA | AK292228.1 | - | 2 - 3 | 1105 | 95 | |
AK301713.1 | - | 3 - 4 | 897 | 95 | ||
AK302301.1 | - | 2 - 3 | 501 | 95 | ||
BC034501.2 | - | 2 - 3 | 1110 | 4823304 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons |