Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001184879.1 | NP_001171808.1 | 6 - 7 | 986 | 73 | |
NM_001184882.1 | NP_001171811.1 | 4 - 5 | 593 | 73 | ||
NM_003874.3 | NP_003865.1 | 5 - 6 | 935 | 73 | ||
XM_011510094.2 | XP_011508396.1 | 6 - 7 | 949 | 73 | ||
XM_011510095.2 | XP_011508397.1 | 5 - 6 | 625 | 73 | ||
GenBank mRNA | AF054815.1 | - | 5 - 6 | 925 | 73 | |
AF054816.1 | - | 6 - 7 | 976 | 73 | ||
AJ223324.1 | - | 5 - 6 | 900 | 73 | ||
AK296290.1 | - | 4 - 5 | 554 | 73 | ||
AK313496.1 | - | 5 - 6 | 901 | 73 | ||
BC020063.1 | - | 5 - 6 | 880 | 4385965 | 73 | |
CR933666.1 | - | 4 - 5 | 554 | 73 | ||
U82988.1 | - | 5 - 6 | 897 | 73 | ||
Y12632.1 | - | 6 - 7 | 958 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |