Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
ENST00000222212.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
ENST00000222212.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
ENST00000222212.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
ENST00000222212.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
ENST00000222212.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
ENST00000222212.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
ENST00000222212.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
ENST00000222212.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
ENST00000391766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
ENST00000391766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
ENST00000391766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
ENST00000391766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
ENST00000391766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
ENST00000391767.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
ENST00000391767.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
ENST00000391767.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
ENST00000391767.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
ENST00000391767.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
ENST00000391767.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
ENST00000391767.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
ENST00000391767.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
ENST00000391767.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
ENST00000391767.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
ENST00000468076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
ENST00000468076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
ENST00000468076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
NM_031896.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
NM_031896.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
NM_031896.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
NM_031896.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
NM_031896.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
NM_031896.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
NM_031896.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
NM_031896.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008826.hg.1 |
NM_031896.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
ENST00000222212.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
ENST00000222212.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
ENST00000222212.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
ENST00000222212.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
ENST00000222212.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
ENST00000222212.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
ENST00000222212.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
ENST00000222212.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
ENST00000391766.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
ENST00000391766.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
ENST00000391766.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
ENST00000391766.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
ENST00000391766.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
ENST00000391767.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
ENST00000391767.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
ENST00000391767.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
ENST00000391767.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
ENST00000391767.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
ENST00000391767.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
ENST00000391767.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
ENST00000391767.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
ENST00000391767.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
ENST00000391767.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
ENST00000468076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
ENST00000468076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
ENST00000468076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
NM_031896.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
NM_031896.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
NM_031896.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
NM_031896.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
NM_031896.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
NM_031896.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
NM_031896.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
NM_031896.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008826.hg.1 |
NM_031896.4 |
Refseq |