Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000588292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000588292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000588292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000588430.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000588430.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000588430.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000588430.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000588430.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000588430.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000588430.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
ENST00000588430.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006539.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000615.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000615.hg.1 |
TC19000042.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000616.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000616.hg.1 |
TC19000042.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900145441.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145441.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145441.hg.1 |
ENST00000588430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145441.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900145441.hg.1 |
TC1900006539.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900145441.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145441.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145441.hg.1 |
ENST00000588430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145441.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000588292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000588292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000588292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000588430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000588430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000588430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000588430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000588430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000588430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000588430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
ENST00000588430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006539.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900145442.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145442.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145442.hg.1 |
ENST00000588430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145442.hg.1 |
NM_001243079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900145442.hg.1 |
TC1900006539.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900145442.hg.1 |
ENST00000334770.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145442.hg.1 |
ENST00000454744.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145442.hg.1 |
ENST00000588430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145442.hg.1 |
NM_001243079.1 |
Refseq |