Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAS1R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138697.3 | NP_619642.2 | 3 - 4 | 1354 | 101 | |
XM_011542203.2 | XP_011540505.1 | 3 - 4 | 1720 | 101 | ||
XM_011542206.2 | XP_011540508.1 | 2 - 3 | 1906 | 101 | ||
XM_017002402.1 | XP_016857891.1 | 2 - 3 | 1906 | 101 | ||
GenBank mRNA | AF387618.1 | - | 2 - 3 | 1067 | 101 | |
AK315805.1 | - | 3 - 4 | 1323 | 101 | ||
BC136515.1 | - | 3 - 4 | 1354 | 9020505 | 101 | |
BC136516.1 | - | 3 - 4 | 1353 | 9020506 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Ion Channel Mammalian Gene Collection Homo sapiens Probe spans exons |