Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014032.hg.1 |
ENST00000300961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014032.hg.1 |
NM_144616.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014032.hg.1 |
TC19001025.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014031.hg.1 |
ENST00000300961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014031.hg.1 |
NM_144616.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014031.hg.1 |
TC19001025.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900203497.hg.1 |
ENST00000300961.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203497.hg.1 |
NM_144616.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900203497.hg.1 |
TC1900011838.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900203497.hg.1 |
ENST00000300961.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203497.hg.1 |
NM_144616.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900203498.hg.1 |
ENST00000300961.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203498.hg.1 |
NM_144616.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900203498.hg.1 |
TC1900011838.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900203498.hg.1 |
ENST00000300961.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203498.hg.1 |
NM_144616.3 |
Refseq |