Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000562190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000562190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000562190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000562190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000562190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000563996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000563996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000563996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000563996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000563996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000563996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000563996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000564009.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000564009.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000564009.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000564915.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000564915.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000564915.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000564915.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000567417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000567417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000567417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000611270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000611270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000611270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
ENST00000611270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500154394.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500154394.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500154394.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500154394.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500154394.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500154394.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500154394.hg.1 |
TC1500007659.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000249636.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000545237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000562190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000562190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000562190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000562190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000562190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000563996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000563996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000563996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000563996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000563996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000563996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000563996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000564009.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000564009.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000564009.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000564915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000564915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000564915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000564915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000567417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000567417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000567417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000611270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000611270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000611270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
ENST00000611270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007659.hg.1 |
NM_016166.2 |
Refseq |