Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05023952.hg.1 |
TC05001769.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05023952.hg.1 |
ENST00000231454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05023952.hg.1 |
NM_000879.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05023954.hg.1 |
TC05001769.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05023954.hg.1 |
ENST00000231454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05023954.hg.1 |
NM_000879.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012019.hg.1 |
ENST00000231454.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012019.hg.1 |
ENST00000231454.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012019.hg.1 |
ENST00000231454.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012019.hg.1 |
ENST00000231454.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012019.hg.1 |
ENST00000231454.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012019.hg.1 |
ENST00000231454.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012019.hg.1 |
ENST00000231454.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012019.hg.1 |
ENST00000450655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012019.hg.1 |
ENST00000450655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012019.hg.1 |
ENST00000450655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012019.hg.1 |
ENST00000462418.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012019.hg.1 |
ENST00000462418.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012019.hg.1 |
ENST00000462418.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012019.hg.1 |
ENST00000462418.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012019.hg.1 |
ENST00000462418.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012019.hg.1 |
NM_000879.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012019.hg.1 |
NM_000879.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012019.hg.1 |
NM_000879.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012019.hg.1 |
NM_000879.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012019.hg.1 |
NM_000879.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012019.hg.1 |
NM_000879.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012019.hg.1 |
NM_000879.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500182955.hg.1 |
ENST00000231454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500182955.hg.1 |
ENST00000462418.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500182955.hg.1 |
NM_000879.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500182955.hg.1 |
ENST00000231454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500182955.hg.1 |
ENST00000462418.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500182955.hg.1 |
NM_000879.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500182955.hg.1 |
TC0500012019.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500182952.hg.1 |
ENST00000231454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500182952.hg.1 |
ENST00000450655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500182952.hg.1 |
ENST00000462418.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500182952.hg.1 |
NM_000879.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500182952.hg.1 |
ENST00000231454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500182952.hg.1 |
ENST00000450655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500182952.hg.1 |
ENST00000462418.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500182952.hg.1 |
NM_000879.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500182952.hg.1 |
TC0500012019.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500012019.hg.1 |
ENST00000231454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012019.hg.1 |
ENST00000231454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012019.hg.1 |
ENST00000231454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012019.hg.1 |
ENST00000231454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012019.hg.1 |
ENST00000231454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012019.hg.1 |
ENST00000231454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012019.hg.1 |
ENST00000231454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012019.hg.1 |
ENST00000450655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012019.hg.1 |
ENST00000450655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012019.hg.1 |
ENST00000450655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012019.hg.1 |
ENST00000462418.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012019.hg.1 |
ENST00000462418.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012019.hg.1 |
ENST00000462418.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012019.hg.1 |
ENST00000462418.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012019.hg.1 |
ENST00000462418.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012019.hg.1 |
NM_000879.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012019.hg.1 |
NM_000879.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012019.hg.1 |
NM_000879.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012019.hg.1 |
NM_000879.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012019.hg.1 |
NM_000879.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012019.hg.1 |
NM_000879.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012019.hg.1 |
NM_000879.2 |
Refseq |